2-(Tributylstannyl)thiazole ≥95%

Supplier: Matrix Scientific

007396-5G 007396-1G
101779-556EA 715.38 USD
101779-556 101779-554
2-(Tributylstannyl)thiazole ≥95%
2-(Tributylstannyl)thiazole

Hazard: Toxic/Irritant/Refrigerate

Formula: C₁₅H₂₉NSSn
Boiling Pt: 307-308°C
Melting Pt: 224-225°C
Density: 1.19
MDL Number: MFCD01319057
CAS Number: 121359-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR