3-(Tributylstannyl)pyridine ≥93%

Supplier: Matrix Scientific

Synonyms: 3-(Tri-n-butylstannyl)pyridine

007393-1G 007393-5G
101779-546EA 29.23 USD
101779-546 101779-548
3-(Tributylstannyl)pyridine ≥93%
3-(Tributylstannyl)pyridine

Hazard: Toxic/Irritant/Refrigerate

Formula: C₁₇H₃₁NSn
Density: 1.153
MDL Number: MFCD00052054
CAS Number: 59020-10-9
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR