2,4,5-Trichlorobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4,5-Trichlorobenzene-1-sulphonyl chloride

007250-25G 007250-5G
101779-534EA 130.77 USD
101779-534 101779-532
2,4,5-Trichlorobenzenesulfonyl chloride ≥98%
2,4,5-Trichlorobenzenesulphonyl chloride

TCA listed: T
Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₂Cl₄O₂S
MW: 279.96 g/mol
Melting Pt: 68…70 °C
MDL Number: MFCD00007428
CAS Number: 15945-07-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 68-70°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR