2-Methylallylamine

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-methyl-1-propene

007222-5G 007222-1G 007222-25G
101779-508EA 105.92 USD
101779-508 101779-506 101779-510
2-Methylallylamine
2-Methylallylamine

Hazard: Flammable/Corrosive

Formula: C₄H₉N
MW: 71.123 g/mol
Boiling Pt: 78°C
Density: 0.78
MDL Number: MFCD00053646
CAS Number: 2878-14-0

Order Now

Specification Test Results

Boiling point 78°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR