2,2-Difluorobutyric acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2-Difluorobutanoic acid

007217-250MG 007217-1G
101779-502EA 43.08 USD
101779-502 101779-504
2,2-Difluorobutyric acid ≥97%
2,2-Difluorobutyric acid

Hazard: Corrosive/Irritant

Formula: C₄H₆F₂O₂
Boiling Pt: 141 °C
Melting Pt: 7…9 °C
Density: 1.22
MDL Number: MFCD03093765
CAS Number: 2366-62-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 141°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR