2,2-Difluoropropionic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2-Difluoropropanoic acid

007216-1G 007216-5G
101779-496EA 30.77 USD
101779-496 101779-498
2,2-Difluoropropionic acid ≥97%
2,2-Difluoropropionic acid

Hazard: Corrosive/Moisture Sensitive

Formula: C₃H₄F₂O₂
Boiling Pt: 142 °C
Melting Pt: 38…45 °C
MDL Number: MFCD03093764
CAS Number: 373-96-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 142°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR