5-Fluoro-o-cresol
Supplier: Matrix Scientific
Synonyms:
5-Fluoro-2-methylphenol, 1-Fluoro-3-hydroxy-4-methylbenzene
007206-5G
007206-25G
101779-484EA
67.69
USD
101779-484
101779-486
5-Fluoro-o-cresol
5-Fluoro-o-cresol
Hazard: Corrosive/Irritant
Formula:
C₇H₇FO MW: 126.13 g/mol Boiling Pt: 66 °C (4 mmHg) Density: 1.157 |
MDL Number:
MFCD00673009 CAS Number: 452-85-7 UN: 3265 ADR: 8,III |
Specification Test Results
Boiling point | 66°/4mm |
Density | 1.157 |
ND20 | 1.514 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...