3,5-Difluorobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5-Difluorobenzene-1-sulphonyl chloride

007204-5G 007204-100G 007204-25G
101779-478EA 20 USD
101779-478 101779-482 101779-480
3,5-Difluorobenzenesulfonyl chloride ≥98%
3,5-Difluorobenzenesulphonyl chloride

Hazard: Corrosive

Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Melting Pt: 57…59 °C
MDL Number: MFCD02091378
CAS Number: 210532-25-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 57-59°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR