3-Cyano-4-fluorobenzenesulfonyl chloride

Supplier: Matrix Scientific

007192-5G 007192-25G
101779-474EA 146.15 USD
101779-474 101779-476
3-Cyano-4-fluorobenzenesulfonyl chloride
3-Cyano-4-fluorobenzenesulphonyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₇H₃ClFNO₂S
Melting Pt: 67-70°C
MDL Number: MFCD03093760
CAS Number: 351003-23-1
UN: 3261
ADR: 8,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR