2,4-Dibromobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4-Dibromobenzene-1-sulphonyl chloride

007191-1G 007191-5G
101779-470EA 33.85 USD
101779-470 101779-472
2,4-Dibromobenzenesulfonyl chloride ≥98%
2,4-Dibromobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₃Br₂ClO₂S
MW: 334.42 g/mol
Boiling Pt: 128 °C (0.2 mmHg)
Melting Pt: 82…86 °C
MDL Number: MFCD03093759
CAS Number: 72256-95-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 128°/0.2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR