2-Amino-6-fluorobenzylamine

Supplier: Matrix Scientific

007187-1G 007187-5G
101779-466EA 38.46 USD
101779-466 101779-468
2-Amino-6-fluorobenzylamine
2-Amino-6-fluorobenzylamine

Hazard: Corrosive

Formula: C₇H₉FN₂
Boiling Pt: 78-80°C/1mm
MDL Number: MFCD00042483
CAS Number: 175277-93-7

Order Now

Specification Test Results

Boiling point 78-80°/1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR