2-Chloro-6-(trifluoromethyl)pyridine ≥98%
Supplier: Matrix Scientific
007184-25G
007184-100G
101779-458EA
30.77
USD
101779-458
101779-460
2-Chloro-6-(trifluoromethyl)pyridine ≥98%
2-Chloro-6-(trifluoromethyl)pyridine
Hazard: Toxic
Formula:
C₆H₃ClF₃N Boiling Pt: 60…62 °C (8 mmHg) Melting Pt: 28…29 °C |
MDL Number:
MFCD00042226 CAS Number: 39890-95-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Boiling point | 60-62°/8mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...