2,3,3,3-Tetrafluoropropionic acid

Supplier: Matrix Scientific

007180-5G 007180-25G
101779-454EA 118.46 USD
101779-454 101779-456
2,3,3,3-Tetrafluoropropionic acid
2,3,3,3-Tetrafluoropropionic acid

Hazard: Corrosive/Hygroscopic

Formula: C₃H₂F₄O₂
Boiling Pt: 120…121 °C
Melting Pt: 28…29 °C
Density: 1.54
MDL Number: MFCD01328323
CAS Number: 359-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR