(+/-)-α-Methoxy-α-trifluoromethylphenylacetyl chloride ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-2-phenyl-3,3,3-trifluoropropanoyl chloride, (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

007172-1G 007172-5G
101779-450EA 38.46 USD
101779-450 101779-452
(+/-)-α-Methoxy-α-trifluoromethylphenylacetyl chloride ≥99%
(+/-)-alpha-Methoxy-alpha-trifluoromethylphenylacetyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₁₀H₈ClF₃O₂
MW: 252.62 g/mol
Boiling Pt: 213…214 °C
MDL Number: MFCD00078209
CAS Number: 40793-68-8

Order Now

Specification Test Results

Boiling point 213-214°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR