(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99%

Supplier: Matrix Scientific

Synonyms: (+)-Mosher's Acid Chloride, (+)-MTPA-Cl, (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, (S)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoyl chloride

007170-250MG 007170-1G
101779-446EA 98.46 USD
101779-446 101779-448
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99%
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₁₀H₈ClF₃O₂
MW: 252.62 g/mol
Boiling Pt: 213…214 °C
Density: 1.353
MDL Number: MFCD00067105
CAS Number: 20445-33-4

Order Now

Specification Test Results

Boiling point 213-214°
Density 1.353
ND20 1.469

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR