(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99%

Supplier: Matrix Scientific

Synonyms: (-)-MTPA-Cl, (-)-Mosher's Acid Chloride, (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

007169-1G 007169-250MG
101779-444EA 330.77 USD
101779-444 101779-442
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive/Refrigerate

Formula: C₁₀H₈ClF₃O₂
MW: 252.62 g/mol
Boiling Pt: 213…214 °C
Density: 1.353
MDL Number: MFCD00044400
CAS Number: 39637-99-5

Order Now

Specification Test Results

Boiling point 213-214°
Density 1.353
ND20 1.469

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR