4,4,4-Trifluorobutyronitrile ≥97%

Supplier: Matrix Scientific

007160-1G 007160-5G
101779-432EA 149.23 USD
101779-432 101779-434
4,4,4-Trifluorobutyronitrile ≥97%
4,4,4-Trifluorobutyronitrile

Hazard: Flammable/Toxic

Formula: C₄H₄F₃N
MW: 123.08 g/mol
Boiling Pt: 138-140°C
MDL Number: MFCD02183570
CAS Number: 690-95-9

Order Now

Specification Test Results

Boiling point 138-140°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR