2,5-Dichloro-3-thiophenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
007141-1G
007141-5G
101779-400EA
75.38
USD
101779-400
101779-402
2,5-Dichloro-3-thiophenesulfonyl chloride ≥97%
2,5-Dichloro-3-thiophenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₄HCl₃O₂S₂ MW: 251.54 Boiling Pt: 105°C/2mm |
MDL Number:
MFCD00051665 CAS Number: 56946-83-9 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 105°/2mm |
ND20 | 1.597 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...