3-Cyanobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Cyanobenzene-1-sulphonyl chloride, 3-Cyanobenzene-1-sulphonylchloride

007139-1G 007139-25G 007139-5G
101779-390EA 21.54 USD
101779-390 101779-394 101779-392
3-Cyanobenzenesulfonyl chloride ≥98%
3-Cyanobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
Boiling Pt: 114…116 °C (0.3 mmHg)
Melting Pt: 46…47 °C
MDL Number: MFCD00203480
CAS Number: 56542-67-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 114-116°/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR