2-Cyanobenzenesulphonyl chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Cyanobenzene-1-sulphonyl chloride
007138-25G
007138-5G
101779-388EA
113.85
USD
101779-388
101779-386
2-Cyanobenzenesulphonyl chloride ≥95%
2-Cyanobenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₇H₄ClNO₂S MW: 201.63 Melting Pt: 67.5 |
MDL Number:
MFCD00052341 CAS Number: 69360-26-5 UN: 3261 ADR: 8 II |
Specification Test Results
Melting point | 66-68° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...