3-Chloro-4-methylbenzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Chlor-4-methylbenzenesulphonyl chloride, 2-Chloro-p-toluenesulphochloride, 3-Chloro-4-methylbenzene-1-sulphonyl chloride
007135-5G
007135-25G
101779-378EA
20
USD
101779-378
101779-380
3-Chloro-4-methylbenzenesulfonyl chloride ≥97%
3-Chloro-4-methylbenzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₇H₆Cl₂O₂S MW: 225.09 g/mol Boiling Pt: 94-96°C/0.05mm |
MDL Number:
MFCD00203987 CAS Number: 42413-03-6 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 94-96°/0.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...