4-(Bromomethyl)benzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(Bromomethyl)benzene-1-sulphonyl chloride
007130-100G
007130-5G
007130-25G
101779-366EA
600
USD
101779-366
101779-362
101779-364
4-(Bromomethyl)benzenesulfonyl chloride ≥97%
4-(Bromomethyl)benzenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₇H₆BrClO₂S MW: 269.55 g/mol Melting Pt: 77…79 °C |
MDL Number:
MFCD00156129 CAS Number: 66176-39-4 UN: 3261 ADR: 8,II |
Specification Test Results
Melting point | 77-79° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...