3-Bromo-5-chloro-2-thiophenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
007128-1G
007128-250MG
101779-356EA
773.85
USD
101779-356
101779-354
3-Bromo-5-chloro-2-thiophenesulfonyl chloride ≥97%
3-Bromo-5-chloro-2-thiophenesulphonyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₄HBrCl₂O₂S₂ Melting Pt: 34…36 °C |
MDL Number:
MFCD00052095 CAS Number: 175205-72-8 |
Specification Test Results
Melting point | 34-36° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...