3-(Trifluoromethoxy)benzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

007092-25G 007092-1G 007092-5G
101779-340EA 215.38 USD
101779-340 101779-336 101779-338
3-(Trifluoromethoxy)benzenesulfonyl chloride ≥98%
3-(Trifluoromethoxy)benzenesulphonyl chloride

Hazard: Corrosive

Formula: C₇H₄ClF₃O₃S
Boiling Pt: 73…74 °C (0.7 mmHg)
MDL Number: MFCD01091016
CAS Number: 220227-84-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 73-74°/0.7
ND20 1.4754


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR