4-Chloro-3-picoline hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-methylpyridine hydrochloride

007068-1G 007068-5G
101779-332EA 29.23 USD
101779-332 101779-334
4-Chloro-3-picoline hydrochloride ≥97%
4-Chloro-3-picoline hydrochloride

Hazard: Corrosive/Irritant/Refrigerate

Formula: C₆H₇Cl₂N
MW: 164.04 g/mol
Boiling Pt: 76-78°C/2.4mm
Melting Pt: 166-170°C
MDL Number: MFCD03092890
CAS Number: 19524-08-4

Order Now

Specification Test Results

Boiling point 76-78°/2.4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR