6-Bromo-1-hexene ≥97%

Supplier: Matrix Scientific

007061-25G 007061-5G
101779-330EA 89.23 USD
101779-330 101779-328
6-Bromo-1-hexene ≥97%
6-Bromo-1-hexene

TCA listed: T
Hazard: Flammable

Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 48-49°C/16mm
Density: 1.217
MDL Number: MFCD00000269
CAS Number: 2695-47-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 48-49°/16m
Density 1.217
ND20 1.4652

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR