2-Amino-4,5-dimethoxybenzonitrile ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-4,5-dimethoxyaniline

007054-25G 007054-5G
101779-324EA 638.46 USD
101779-324 101779-322
2-Amino-4,5-dimethoxybenzonitrile ≥99%
2-Amino-4,5-dimethoxybenzonitrile

Hazard: Toxic

Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
Melting Pt: 97-101°C
MDL Number: MFCD00058682
CAS Number: 26961-27-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 98-99°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR