3,5-Dichlorobenzoyl chloride ≥97%
Supplier: Matrix Scientific
007011-25G
007011-100G
101779-308EA
36.92
USD
101779-308
102550-892
3,5-Dichlorobenzoyl chloride ≥97%
3,5-Dichlorobenzoyl chloride
TCA listed: T
Hazard: Corrosive/Moisture Sensitive/Refrigerate/Lachrymator
Formula:
C₇H₃Cl₃O Boiling Pt: 135…137 °C (25 mmHg) Melting Pt: 26…28 °C |
MDL Number:
MFCD00009817 CAS Number: 2905-62-6 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 135-137°/2 |
ND20 | 1.582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...