5-Chloro-2-thiophenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

007007-5G 007007-25G
101779-302EA 20 USD
101779-302 101779-304
5-Chloro-2-thiophenesulfonyl chloride ≥97%
5-Chloro-2-thiophenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
Boiling Pt: 117…120 °C (14 mmHg)
Melting Pt: 25…28 °C
Density: 1.623
MDL Number: MFCD00051667
CAS Number: 2766-74-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 117-120°/1
Density 1.623

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR