2-Chloro-3-nitro-5-thiophenesulfonyl chloride

Supplier: Matrix Scientific

Synonyms: 5-Chloro-4-nitrothiophene-2-sulphonyl chloride

007003-1G 007003-5G
101779-298EA 36.92 USD
101779-298 101779-300
2-Chloro-3-nitro-5-thiophenesulfonyl chloride
2-Chloro-3-nitro-5-thiophenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₄HCl₂NO₄S₂
MW: 262.09 g/mol
Melting Pt: 52-54°C
MDL Number: MFCD00067978
CAS Number: 58457-24-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Melting point 52-54°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR