Coumarin-6-sulfonyl chloride
Supplier: Matrix Scientific
006872-1G
006872-5G
101779-294EA
52.31
USD
101779-294
101779-296
Coumarin-6-sulfonyl chloride
Coumarin-6-sulfonyl chloride
Hazard: Corrosive
Formula:
C₉H₅ClO₄S Melting Pt: 110…113 °C |
MDL Number:
MFCD01941320 CAS Number: 10543-42-7 |
Specification Test Results
Melting point | 110-113° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...