2-(Methylsulfonyl)benzenesulfonyl chloride

Supplier: Matrix Scientific

006840-5G 006840-1G
101779-288EA 340 USD
101779-288 101779-286
2-(Methylsulfonyl)benzenesulfonyl chloride
2-(Methylsulfonyl)benzenesulfonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₇ClO₄S₂
Melting Pt: 136…137 °C
MDL Number: MFCD00216493
CAS Number: 89265-35-0

Order Now

Specification Test Results

Melting point 136-137°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR