ω-Bromo-3-fluoroacetophenone ≥95%
Supplier: Matrix Scientific
Synonyms:
Bromomethyl m-fluorophenyl ketone, 3-Fluorophenacyl bromide, 2-Bromo-1-(3-fluorophenyl)ethan-1-one, α-Bromo-3-fluoroacetophenone, 2-Bromo-3'-fluoroacetophenone
006750-1G
006750-5G
006750-25G
101779-276EA
44.62
USD
101779-276
101779-278
101779-280
ω-Bromo-3-fluoroacetophenone ≥95%
ω-Bromo-3-fluoroacetophenone
Hazard: Corrosive/Refrigerate/Lachrymator
Formula:
C₈H₆BrFO MW: 217.04 g/mol Boiling Pt: 76 °C (0.09 mmHg) Melting Pt: 32…37 °C |
MDL Number:
MFCD00109756 CAS Number: 53631-18-8 |
Specification Test Results
Boiling point | 76°/0.09mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...