2,4,6-Trichlorobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Trichlorobenzene-1-sulphonyl chloride

006737-5G 006737-25G
101779-258EA 38.46 USD
101779-258 101779-260
2,4,6-Trichlorobenzenesulfonyl chloride ≥98%
2,4,6-Trichlorobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₂Cl₄O₂S
MW: 279.96 g/mol
Melting Pt: 43-46°C
MDL Number: MFCD00052973
CAS Number: 51527-73-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 43-46°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR