1-Naphthoyl chloride ≥98%
Supplier: Matrix Scientific
006736-25G
006736-100G
101779-254EA
52.31
USD
101779-254
101779-256
1-Naphthoyl chloride ≥98%
1-Naphthoyl chloride
Hazard: Corrosive/Moisture Sensitive
Formula:
C₁₁H₇ClO MW: 190.63 g/mol Boiling Pt: 118 °C (0.1 mmHg) Melting Pt: 16…19 °C Density: 1.265 |
MDL Number:
MFCD00004002 CAS Number: 879-18-5 UN: 3265 ADR: 8 II |
Specification Test Results
Boiling point | 118°/0.1mm |
Density | 1.265 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...