Methyl adipoyl chloride ≥96%

Supplier: Matrix Scientific

Synonyms: Methyl 6-chloro-6-oxohexanoate

006733-5G 006733-25G
101779-250EA 72.31 USD
101779-250 101779-252
Methyl adipoyl chloride ≥96%
Methyl adipoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₁₁ClO₃
MW: 178.62 g/mol
Boiling Pt: 76 °C (0.8 mmHg)
Density: 1.05
MDL Number: MFCD00013661
CAS Number: 35444-44-1

Order Now

Specification Test Results

Boiling point 76°/0.8mm
Density 1.05
ND20 1.447

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR