3-Butyne-2-one ≥95%
Supplier: Matrix Scientific
Synonyms:
But-3-yn-2-one
006723-5G
006723-25G
101779-242EA
120
USD
101779-242
101779-244
3-Butyne-2-one ≥95%
3-Butyne-2-one
Hazard: Flammable/Toxic/Refrigerate
Formula:
C₄H₄O MW: 68.08 g/mol Boiling Pt: 84-86°C Density: 0.87 Flash Pt: -1°C |
MDL Number:
MFCD00008775 CAS Number: 1423-60-5 UN: 1992 ADR: 3,II |
Specification Test Results
Boiling point | 84-86° |
Density | 0.87 |
ND20 | 1.406 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...