2-(Methylthio)ethylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Methylthio)ethanamine
006716-1G
006716-5G
101779-234EA
55.38
USD
101779-234
101779-236
2-(Methylthio)ethylamine ≥95%
2-(Methylthio)ethylamine
TCA listed: T
Hazard: Flammable/Corrosive/Refrigerate/Stench
Formula:
C₃H₉NS MW: 91.18 g/mol Boiling Pt: 146-148°C Density: 0.974 |
MDL Number:
MFCD00014825 CAS Number: 18542-42-2 |
Specification Test Results
Boiling point | 146-148° |
Density | 0.974 |
ND20 | 1.495 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...