4-Isopropylphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

006646-1G 006646-5G
101779-206EA 25.99 USD
101779-206 101779-208
4-Isopropylphenyl isothiocyanate ≥98%
4-Isopropylphenyl isothiocyanate

Hazard: Toxic/Corrosive/Moisture Sensitive/Lachrymator

Formula: C₁₀H₁₁NS
MW: 177.27 g/mol
Boiling Pt: 155°C/20mm
Density: 1.03
MDL Number: MFCD00022060
CAS Number: 89007-45-4

Order Now

Specification Test Results

Boiling point 155°/20mm
Density 1.03

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR