1,4-Divinyloctafluorobutane ≥97%

Supplier: Matrix Scientific

Synonyms: 1H,1H,2H,7H,8H,8H-Perfluoroocta-1,7-diene

006643-1G 006643-5G
101779-196EA 110.77 USD
101779-196 101779-198
1,4-Divinyloctafluorobutane ≥97%
1,4-Divinyloctafluorobutane

Hazard: Flammable

Formula: C₈H₆F₈
MW: 254.12 g/mol
Boiling Pt: 123 °C
Density: 1.361
MDL Number: MFCD00077520
CAS Number: 678-65-9

Order Now

Specification Test Results

Boiling point 123°
Density 1.361


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR