(Bromomethyl)cyclopropane ≥96%

Supplier: Matrix Scientific

006642-25G 006642-100G
101779-192EA 36.92 USD
101779-192 101779-194
(Bromomethyl)cyclopropane ≥96%
(Bromomethyl)cyclopropane

Hazard: Flammable/Irritant/Refrigerate

Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 105-107°C
Density: 1.392
MDL Number: MFCD00001306
CAS Number: 7051-34-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 105-107°
Density 1.392
ND20 1.457

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR