4-Isopropylbenzenesulphonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Methylethyl)benzolsulphonsäurechlorid, 4-Isopropylbenzene-1-sulphonyl chloride

006626-25G 006626-100G
101779-188EA 60 USD
101779-188 101779-190
4-Isopropylbenzenesulphonyl chloride ≥95%
4-Isopropylbenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₉H₁₁ClO₂S
MW: 218.7
Boiling Pt: 142 °C (5 mmHg)
Density: 1.22
MDL Number: MFCD00041510
CAS Number: 54997-90-9
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR