Cyclopropylamine ≥97%

Supplier: Matrix Scientific

Synonyms: Aminocyclopropane

006620-100G 006620-500G
101779-184EA 69.23 USD
101779-184 101779-186
Cyclopropylamine ≥97%
Cyclopropylamine

TCA listed: T
Hazard: Flammable/Corrosive

Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 48-50°C
Density: 0.824
MDL Number: MFCD00001301
CAS Number: 765-30-0
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Boiling point 48-50°
Density 0.824
ND20 1.4206

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR