Cyclopropylacetic acid ≥96%

Supplier: Matrix Scientific

006618-5G 006618-25G
101779-176EA 87.69 USD
101779-176 101779-178
Cyclopropylacetic acid ≥96%
Cyclopropylacetic acid

Hazard: Corrosive/Stench

Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 88°C/5mm
MDL Number: MFCD00041544
CAS Number: 5239-82-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 88°/5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR