Cyclopentyl isothiocyanate ≥98%

Supplier: Matrix Scientific

006597-1G 006597-5G
101779-170EA 69.23 USD
101779-170 101779-172
Cyclopentyl isothiocyanate ≥98%
Cyclopentyl isothiocyanate

Hazard: Toxic/Moisture Sensitive/Lachrymator

Formula: C₆H₉NS
Boiling Pt: 76 °C (10 mmHg)
Density: 1.03
MDL Number: MFCD00040877
CAS Number: 33522-03-1
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Boiling point 76°/10mm
Density 1.03


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR