Allyl ethyl ether ≥96%

Supplier: Matrix Scientific

Synonyms: Allyl Ethyl Ether

006595-5G 006595-25G
101779-166EA 116.92 USD
101779-166 101779-168
Allyl ethyl ether ≥96%
Allyl ethyl ether

TCA listed: T
Hazard: Flammable/Toxic

Formula: C₅H₁₀O
Boiling Pt: 64…65 °C
Density: 0.8664
MDL Number: MFCD00009255
CAS Number: 557-31-3
UN: 2335
ADR: 3,II

Order Now

Specification Test Results

Boiling point 64-65°
Density 0.76
ND20 1.3822


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR