Sodium hexafluorophosphate ≥99%
Supplier: Matrix Scientific
006532-500G
006532-25G
006532-100G
101779-158EA
638.46
USD
101779-158
101779-154
101779-156
Sodium hexafluorophosphate ≥99%
Sodium hexafluorophosphate
Hazard: Corrosive/Irritant/Hygroscopic
Formula:
F₆NaP Melting Pt: Density: 2.369 |
MDL Number:
MFCD00011122 CAS Number: 21324-39-0 UN: 3260 ADR: 8,II |
Specification Test Results
Density | 2.369 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...