Tetrabutylammonium dihydrogentrifluoride
Supplier: Matrix Scientific
006519-5G
006519-25G
101779-150EA
38.46
USD
101779-150
101779-152
Tetrabutylammonium dihydrogentrifluoride
Tetrabutylammonium dihydrogentrifluoride
Hazard: Flammable/Corrosive
Formula:
C₁₆H₃₈F₃N Flash Pt: 13 °C |
MDL Number:
MFCD00145365 CAS Number: 99337-56-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...