4'-tert-Butylacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: p-tert-Butylacetophenone

006514-25G 006514-100G
101779-142EA 146.15 USD
101779-142 101779-144
4'-tert-Butylacetophenone ≥95%
4'-tert-Butylacetophenone

TCA listed: T
Hazard: Flammable/Refrigerate

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Boiling Pt: 107…108 °C (5 mmHg)
Melting Pt: 17…18 °C
Density: 0.964
Flash Pt: 30 °C
MDL Number: MFCD00017256
CAS Number: 943-27-1

Order Now

Specification Test Results

Boiling point 107-108°/5
Density 0.964
ND20 1.52

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR