3,3'-Difluorobenzophenone ≥98%
Supplier: Matrix Scientific
Synonyms:
Bis(3-fluorophenyl)methanone, Bis(3-fluorophenyl) ketone
006465-1G
006465-5G
101779-106EA
56.92
USD
101779-106
101779-108
3,3'-Difluorobenzophenone ≥98%
3,3'-Difluorobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈F₂O MW: 218.2 g/mol Melting Pt: 59…61 °C |
MDL Number:
MFCD00000337 CAS Number: 345-70-0 |
Specification Test Results
Melting point | 59-61° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...